Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTCTGCCCTGCTGACAAGTCTTG[A/T]CAAGCTAAAAATAAAGTTTATAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600298 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LMX1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LMX1A - LIM homeobox transcription factor 1 alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001174069.1 | 1579 | UTR 3 | NP_001167540.1 | |||
NM_177398.3 | 1579 | UTR 3 | NP_796372.1 | |||
XM_011509538.2 | 1579 | UTR 3 | XP_011507840.1 | |||
XM_011509540.2 | 1579 | Intron | XP_011507842.1 |