Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGTACAGGATACACATTCTTACC[A/G]TGTACACATGGAGCATTTGTCAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602171 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DUSP27 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DUSP27 - dual specificity phosphatase 27 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080426.1 | Intron | NP_001073895.1 | ||||
XM_005245603.3 | Intron | XP_005245660.1 | ||||
XM_011510146.2 | Intron | XP_011508448.1 |
GPA33 - glycoprotein A33 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105371598 - uncharacterized LOC105371598 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |