Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCACCTTCGCCCGTCCCTGTCAC[A/G]TCTAGCGGGCAGCCCTCCTTGTTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
25 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613181 MIM: 185860 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BOLA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BOLA1 - bolA family member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SV2A - synaptic vesicle glycoprotein 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278719.1 | 2035 | Silent Mutation | GAA,GAG | E,E 38 | NP_001265648.1 | |
NM_014849.4 | 2035 | Silent Mutation | GAA,GAG | E,E 586 | NP_055664.3 | |
XM_017002992.1 | 2035 | Silent Mutation | GAA,GAG | E,E 586 | XP_016858481.1 |