Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATTAAGGATTTCCCAAATATATA[A/G]TTTTATCATGATTTATTTCATTTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 134370 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CFH - complement factor H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000186.3 | Intron | NP_000177.2 | ||||
NM_001014975.2 | Intron | NP_001014975.1 | ||||
XM_017001108.1 | Intron | XP_016856597.1 |