Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAAGGCTGTATCAGCCTTCTGCAC[A/G]CATGTTAAGTTCTTATTAAATCAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
27 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613775 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAMSAP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CAMSAP2 - calmodulin regulated spectrin associated protein family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297707.1 | Intron | NP_001284636.1 | ||||
NM_001297708.1 | Intron | NP_001284637.1 | ||||
NM_203459.2 | Intron | NP_982284.1 | ||||
XM_005245041.3 | Intron | XP_005245098.1 | ||||
XM_017000799.1 | Intron | XP_016856288.1 |