Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGGAGCCCTGGTGTTGGTCGAGG[C/T]TGCCCAGGGAGGACCCGAGGTTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
24 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114208 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CACNA1S PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CACNA1S - calcium voltage-gated channel subunit alpha1 S | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000069.2 | 5797 | Missense Mutation | AAC,AGC | N,S 1857 | NP_000060.2 | |
XM_005245478.3 | 5797 | Missense Mutation | AAC,AGC | N,S 1838 | XP_005245535.1 |
LOC101929305 - uncharacterized LOC101929305 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |