Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAATTCATCCCCACACTTAAATA[A/G]GTTCAAAACTAAGAGGTCACCTATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
27 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613257 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP1R15B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPP1R15B - protein phosphatase 1 regulatory subunit 15B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032833.4 | 3932 | UTR 3 | NP_116222.4 | |||
XM_005245551.4 | 3932 | Intron | XP_005245608.2 |