Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTCCGTGGCTGGCTGCCTCCTGC[A/G]GATGTACTTTTTCCACTCACTTGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
32 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
OR6K6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OR6K6 - olfactory receptor family 6 subfamily K member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005184.1 | 383 | Missense Mutation | CAG,CGG | Q,R 128 | NP_001005184.1 |
OR6N1 - olfactory receptor family 6 subfamily N member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005185.1 | 383 | Intron | NP_001005185.1 | |||
XM_017000325.1 | 383 | Intron | XP_016855814.1 | |||
XM_017000326.1 | 383 | Intron | XP_016855815.1 | |||
XM_017000327.1 | 383 | Intron | XP_016855816.1 |