Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATGCTGGAGGAAAAGCTGGTGCC[C/T]CTATTGTATGCATGCTGAACCAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM76A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM76A - family with sequence similarity 76 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143912.1 | Intron | NP_001137384.1 | ||||
NM_001143913.1 | Intron | NP_001137385.1 | ||||
NM_001143914.1 | Intron | NP_001137386.1 | ||||
NM_001143915.1 | Intron | NP_001137387.1 | ||||
NM_152660.2 | Intron | NP_689873.1 | ||||
XM_017000543.1 | Intron | XP_016856032.1 |