Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCTCTTGGGAAATCTTGCAGTAC[C/T]TATGGTACCTGGGACCCCAGGGACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603490 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
WNT4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WNT4 - Wnt family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030761.4 | 2738 | UTR 3 | NP_110388.2 | |||
XM_011541597.2 | 2738 | UTR 3 | XP_011539899.1 | |||
XM_011541598.2 | 2738 | UTR 3 | XP_011539900.1 | |||
XM_011541599.1 | 2738 | Intron | XP_011539901.1 |