Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTCTTTATTTTGTAAGAGTTTGT[A/G]TAGAATATGACATTATTCCTTTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613831 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UBR3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UBR3 - ubiquitin protein ligase E3 component n-recognin 3 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172070.3 | Intron | NP_742067.3 | ||||
XM_005246299.4 | Intron | XP_005246356.1 | ||||
XM_005246300.4 | Intron | XP_005246357.1 | ||||
XM_006712268.3 | Intron | XP_006712331.1 | ||||
XM_006712269.3 | Intron | XP_006712332.1 | ||||
XM_006712270.3 | Intron | XP_006712333.2 | ||||
XM_006712271.3 | Intron | XP_006712334.1 | ||||
XM_011510604.2 | Intron | XP_011508906.1 | ||||
XM_017003356.1 | Intron | XP_016858845.1 | ||||
XM_017003357.1 | Intron | XP_016858846.1 | ||||
XM_017003358.1 | Intron | XP_016858847.1 | ||||
XM_017003359.1 | Intron | XP_016858848.1 |