Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATGCCTGGCCAGCTGCTTATACT[C/T]TGCTTTCTCTCAGAAATAATTGTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147556 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITGA6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ITGA6 - integrin subunit alpha 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000210.3 | Intron | NP_000201.2 | ||||
NM_001079818.2 | Intron | NP_001073286.1 | ||||
NM_001316306.1 | Intron | NP_001303235.1 | ||||
XM_006712510.1 | Intron | XP_006712573.1 | ||||
XM_006712511.1 | Intron | XP_006712574.1 | ||||
XM_017004005.1 | Intron | XP_016859494.1 | ||||
XM_017004006.1 | Intron | XP_016859495.1 | ||||
XM_017004007.1 | Intron | XP_016859496.1 | ||||
XM_017004008.1 | Intron | XP_016859497.1 |