Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGCATTACGATGCTTTGTTCAAA[C/G]ACAGAAATGATACATCATATGGCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614967 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR155 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPR155 - G protein-coupled receptor 155 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033045.3 | 4792 | UTR 3 | NP_001028217.1 | |||
NM_001267050.1 | 4792 | UTR 3 | NP_001253979.1 | |||
NM_001267051.1 | 4792 | UTR 3 | NP_001253980.1 | |||
NM_152529.6 | 4792 | UTR 3 | NP_689742.4 | |||
XM_017003485.1 | 4792 | UTR 3 | XP_016858974.1 | |||
XM_017003486.1 | 4792 | UTR 3 | XP_016858975.1 | |||
XM_017003487.1 | 4792 | UTR 3 | XP_016858976.1 |
SCRN3 - secernin 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |