Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGGCTCCAATGTTGCAGCAGCAG[A/C]AGAACAATTTTGAGCATTCTGGACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606423 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DIRC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DIRC1 - disrupted in renal carcinoma 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052952.2 | 422 | Missense Mutation | GCT,TCT | A,S 51 | NP_443184.1 | |
XM_017003281.1 | 422 | Missense Mutation | GCT,TCT | A,S 51 | XP_016858770.1 |