Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCATCAAGGTACGTACTGCTCCA[A/G]CAGTAACAAGAGCTCCCTATTAATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608307 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CPS1 - carbamoyl-phosphate synthase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122633.2 | Intron | NP_001116105.1 | ||||
NM_001122634.3 | Intron | NP_001116106.1 | ||||
NM_001875.4 | Intron | NP_001866.2 | ||||
XM_011510640.1 | Intron | XP_011508942.1 | ||||
XM_011510641.2 | Intron | XP_011508943.1 | ||||
XM_011510642.2 | Intron | XP_011508944.1 | ||||
XM_011510643.2 | Intron | XP_011508945.1 | ||||
XM_011510644.2 | Intron | XP_011508946.1 |
CPS1-IT1 - CPS1 intronic transcript 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |