Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAAAAAGGGACAGGGGAGGGAAG[C/T]TGGAGCAGAAGGACACCTCCCTAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614184 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DIS3L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DIS3L2 - DIS3 like 3'-5' exoribonuclease 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257281.1 | Intron | NP_001244210.1 | ||||
NM_001257282.1 | Intron | NP_001244211.1 | ||||
NM_152383.4 | Intron | NP_689596.4 |