Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGGATTATAGATGTGTAGAAGTA[C/T]GTTTGTAAAATTTTAAAGTTACCTA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 609381 | |||||||||||||||||||||||
Literature Links: |
STXBP5L PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
STXBP5L - syntaxin binding protein 5 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308330.1 | Intron | NP_001295259.1 | ||||
NM_014980.2 | Intron | NP_055795.1 | ||||
XM_006713825.3 | Intron | XP_006713888.1 | ||||
XM_011513328.2 | Intron | XP_011511630.1 | ||||
XM_011513331.2 | Intron | XP_011511633.1 | ||||
XM_011513332.2 | Intron | XP_011511634.1 | ||||
XM_011513333.2 | Intron | XP_011511635.1 | ||||
XM_017007530.1 | Intron | XP_016863019.1 | ||||
XM_017007531.1 | Intron | XP_016863020.1 | ||||
XM_017007532.1 | Intron | XP_016863021.1 | ||||
XM_017007533.1 | Intron | XP_016863022.1 | ||||
XM_017007534.1 | Intron | XP_016863023.1 | ||||
XM_017007535.1 | Intron | XP_016863024.1 |