Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCCACATATCACAACCCTGGGAA[A/C]CTCTCCAGGATCACCCAGGCTGGGA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
DZIP1L PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
DZIP1L - DAZ interacting zinc finger protein 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170538.1 | 4394 | Intron | NP_001164009.1 | |||
NM_173543.2 | 4394 | UTR 3 | NP_775814.2 | |||
XM_005247198.3 | 4394 | UTR 3 | XP_005247255.1 | |||
XM_005247200.4 | 4394 | Intron | XP_005247257.1 | |||
XM_006713526.1 | 4394 | Intron | XP_006713589.1 | |||
XM_006713527.3 | 4394 | Intron | XP_006713590.1 | |||
XM_011512518.2 | 4394 | Intron | XP_011510820.1 | |||
XM_011512520.2 | 4394 | Intron | XP_011510822.1 | |||
XM_011512521.2 | 4394 | Intron | XP_011510823.1 | |||
XM_011512522.1 | 4394 | Intron | XP_011510824.1 | |||
XM_011512524.2 | 4394 | Intron | XP_011510826.1 | |||
XM_011512528.2 | 4394 | Intron | XP_011510830.1 | |||
XM_017005841.1 | 4394 | Intron | XP_016861330.1 | |||
XM_017005842.1 | 4394 | Intron | XP_016861331.1 |