Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTCTCAGTTTTAGCTGGTTGTTA[C/T]GTAATCAAATATCCCATGCATGTTT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 602382 | |||||||||||||||||||||||
Literature Links: |
PLD1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
PLD1 - phospholipase D1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130081.2 | 4101 | UTR 3 | NP_001123553.1 | |||
NM_002662.4 | 4101 | UTR 3 | NP_002653.1 | |||
XM_005247533.2 | 4101 | UTR 3 | XP_005247590.1 | |||
XM_005247534.2 | 4101 | UTR 3 | XP_005247591.1 | |||
XM_011512897.1 | 4101 | Intron | XP_011511199.1 | |||
XM_011512898.1 | 4101 | Intron | XP_011511200.1 | |||
XM_011512899.2 | 4101 | UTR 3 | XP_011511201.1 | |||
XM_017006623.1 | 4101 | Intron | XP_016862112.1 | |||
XM_017006624.1 | 4101 | UTR 3 | XP_016862113.1 |