Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAAAAATGTTTTTTGAGTACTTGC[C/T]ATGTGGCAGGTATAGGCTCTTGTCA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 604958 | |||||||||||||||||||||||
Literature Links: |
ACTL6A PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ACTL6A - actin like 6A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004301.4 | Intron | NP_004292.1 | ||||
NM_177989.3 | Intron | NP_817126.1 | ||||
NM_178042.3 | Intron | NP_829888.1 |