Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGCTATGGGGAGTGAGTGTGTGGA[C/T]GAACAACAGGAACAGGCAGAGTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614690 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C4orf48 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C4orf48 - chromosome 4 open reading frame 48 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001141936.2 | Intron | NP_001135408.2 | ||||
NM_001168243.1 | Intron | NP_001161715.1 | ||||
XM_011513469.2 | Intron | XP_011511771.1 | ||||
XM_011513471.2 | Intron | XP_011511773.1 | ||||
XM_017008190.1 | Intron | XP_016863679.1 |