Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTAAAATATGATTAATAGTCTAAA[A/C]CCCATTTTAACTTTTTTCTCTCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607819 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC30A5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC30A5 - solute carrier family 30 member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001251969.1 | Intron | NP_001238898.1 | ||||
NM_022902.4 | Intron | NP_075053.2 | ||||
NM_024055.4 | Intron | NP_076960.1 | ||||
XM_005248569.3 | Intron | XP_005248626.1 | ||||
XM_006714672.3 | Intron | XP_006714735.1 | ||||
XM_017009749.1 | Intron | XP_016865238.1 |