Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGAACTTCCATGTGTTATTTCCTT[A/C]TCTTGTTGTTTGTGCCTATTGCATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610291 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SV2C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SV2C - synaptic vesicle glycoprotein 2C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297716.1 | Intron | NP_001284645.1 | ||||
NM_014979.3 | Intron | NP_055794.3 | ||||
XM_011543281.2 | Intron | XP_011541583.1 | ||||
XM_011543282.2 | Intron | XP_011541584.2 | ||||
XM_017009244.1 | Intron | XP_016864733.1 |