Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGCAAAAACCCAAAACAAAAATAC[C/T]TAAAGTTGTTAATTTTTGATAACAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614315 | ||||||||||||||||||||
Literature Links: |
ACOT12 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACOT12 - acyl-CoA thioesterase 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_130767.2 | 1287 | Intron | NP_570123.1 | |||
XM_006714531.2 | 1287 | Intron | XP_006714594.1 | |||
XM_006714532.3 | 1287 | Intron | XP_006714595.1 | |||
XM_017009045.1 | 1287 | Intron | XP_016864534.1 | |||
XM_017009046.1 | 1287 | Intron | XP_016864535.1 | |||
XM_017009047.1 | 1287 | Intron | XP_016864536.1 | |||
XM_017009048.1 | 1287 | Intron | XP_016864537.1 |
ZCCHC9 - zinc finger CCHC-type containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001131035.1 | 1287 | Missense Mutation | CCT,CTT | P,L 266 | NP_001124507.1 | |
NM_001131036.1 | 1287 | Missense Mutation | CCT,CTT | P,L 266 | NP_001124508.1 | |
NM_032280.2 | 1287 | Missense Mutation | CCT,CTT | P,L 266 | NP_115656.1 | |
XM_005248616.3 | 1287 | Missense Mutation | CCT,CTT | P,L 115 | XP_005248673.1 |