Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGATTGTTCAAAATTTCCTTTTC[A/G]ATGTGAGAGAAGGATAGCCGCCATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613973 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MB21D1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MB21D1 - Mab-21 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138441.2 | 1295 | Silent Mutation | ATA,ATG | I,M 382 | NP_612450.2 | |
XM_017010232.1 | 1295 | Silent Mutation | ATA,ATG | I,M 382 | XP_016865721.1 |