Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGTATAATTTTAATAGCAGCTAT[C/T]GTTTTTTCTTTTTTTTTGAGACGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600970 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYO6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYO6 - myosin VI | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300899.1 | Intron | NP_001287828.1 | ||||
NM_004999.3 | Intron | NP_004990.3 | ||||
XM_005248719.3 | Intron | XP_005248776.1 | ||||
XM_005248720.3 | Intron | XP_005248777.1 | ||||
XM_005248721.3 | Intron | XP_005248778.1 | ||||
XM_005248722.3 | Intron | XP_005248779.1 | ||||
XM_005248724.3 | Intron | XP_005248781.1 | ||||
XM_005248726.3 | Intron | XP_005248783.1 | ||||
XM_017010899.1 | Intron | XP_016866388.1 |