Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTGAGCTCCATGGTGGGGTTCTG[C/T]GATTTGTTCTATTCACTATGGTCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608268 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RRAGD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RRAGD - Ras related GTP binding D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021244.4 | 2292 | UTR 3 | NP_067067.1 | |||
XM_005248755.4 | 2292 | UTR 3 | XP_005248812.1 | |||
XM_011536022.2 | 2292 | UTR 3 | XP_011534324.1 |