Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAATTTTTATATCAATGGATTGTC[A/G]CTGTACAAAACTAGGTGAAATACTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613209 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM181 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM181 - transmembrane protein 181 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020823.1 | Intron | NP_065874.1 | ||||
XM_005267074.3 | Intron | XP_005267131.1 | ||||
XM_011535999.1 | Intron | XP_011534301.1 | ||||
XM_011536000.1 | Intron | XP_011534302.1 | ||||
XM_011536001.2 | Intron | XP_011534303.1 | ||||
XM_011536002.2 | Intron | XP_011534304.1 | ||||
XM_011536003.2 | Intron | XP_011534305.1 |