Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGAGGAAAGCCAAAGGAGGAAGT[A/G]TTTTGTAGGGATGAGCGGCTGTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608780 MIM: 614725 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GTF2H5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GTF2H5 - general transcription factor IIH subunit 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207118.2 | Intron | NP_997001.1 | ||||
XM_017010862.1 | Intron | XP_016866351.1 |
SERAC1 - serine active site containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |