Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACAGTTTTTCTTAAATTTTCACT[A/G]TATAACTGGGAGTGAGGAGGGAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 154235 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAK - male germ cell associated kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242385.1 | Intron | NP_001229314.1 | ||||
NM_001242957.2 | Intron | NP_001229886.1 | ||||
NM_005906.5 | Intron | NP_005897.1 | ||||
XM_011514619.2 | Intron | XP_011512921.1 | ||||
XM_011514620.2 | Intron | XP_011512922.1 | ||||
XM_011514621.2 | Intron | XP_011512923.1 | ||||
XM_011514622.2 | Intron | XP_011512924.1 | ||||
XM_011514624.2 | Intron | XP_011512926.1 | ||||
XM_017010863.1 | Intron | XP_016866352.1 | ||||
XM_017010864.1 | Intron | XP_016866353.1 | ||||
XM_017010865.1 | Intron | XP_016866354.1 | ||||
XM_017010866.1 | Intron | XP_016866355.1 |
TMEM14B - transmembrane protein 14B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |