Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCACATTCATTGCACTTATAGGG[C/T]TTCTCCCCAGTGTGAATTCGTTGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602277 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF184 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF184 - zinc finger protein 184 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318891.1 | 2289 | Silent Mutation | AAA,AAG | K,K 668 | NP_001305820.1 | |
NM_001318892.1 | 2289 | Silent Mutation | AAA,AAG | K,K 668 | NP_001305821.1 | |
NM_001318893.1 | 2289 | Silent Mutation | AAA,AAG | K,K 592 | NP_001305822.1 | |
NM_007149.2 | 2289 | Silent Mutation | AAA,AAG | K,K 668 | NP_009080.2 | |
XM_011514861.2 | 2289 | Silent Mutation | AAA,AAG | K,K 592 | XP_011513163.1 |