Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCATGAAGAAAAAATACCTGTCT[C/T]CCAAAACAGAGATAATCATCCAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603978 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZSCAN12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZSCAN12 - zinc finger and SCAN domain containing 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163391.1 | 6194 | Intron | NP_001156863.1 | |||
XM_011515014.2 | 6194 | UTR 3 | XP_011513316.1 | |||
XM_011515015.2 | 6194 | UTR 3 | XP_011513317.1 | |||
XM_011515017.2 | 6194 | UTR 3 | XP_011513319.1 | |||
XM_017011528.1 | 6194 | UTR 3 | XP_016867017.1 |