Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTAATAATACCTTATTAACAAGTA[A/G]CACCTAGAGAGCTAGATTGTGTTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616856 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRPF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BRPF3 - bromodomain and PHD finger containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015695.2 | Intron | NP_056510.2 | ||||
XM_005249010.2 | Intron | XP_005249067.1 | ||||
XM_005249011.3 | Intron | XP_005249068.1 | ||||
XM_011514489.1 | Intron | XP_011512791.1 | ||||
XM_011514490.1 | Intron | XP_011512792.1 | ||||
XM_011514491.1 | Intron | XP_011512793.1 | ||||
XM_011514492.1 | Intron | XP_011512794.1 | ||||
XM_017010742.1 | Intron | XP_016866231.1 | ||||
XM_017010743.1 | Intron | XP_016866232.1 |
LOC102723648 - uncharacterized LOC102723648 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |