Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAATATTCCTGCACTAGTGATGGT[C/G]ACTTAGAATGTTTATAGAAATGTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604786 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARL4A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARL4A - ADP ribosylation factor like GTPase 4A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037164.2 | 2697 | UTR 3 | NP_001032241.1 | |||
NM_001195396.1 | 2697 | UTR 3 | NP_001182325.1 | |||
NM_005738.4 | 2697 | UTR 3 | NP_005729.1 | |||
NM_212460.3 | 2697 | UTR 3 | NP_997625.1 |