Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAAAAATACCCTTGCTGATGTAGC[C/T]AGCGATACACCAGGACAATTTCGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603917 MIM: 614905 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EIF3B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EIF3B - eukaryotic translation initiation factor 3 subunit B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037283.1 | Intron | NP_001032360.1 | ||||
NM_003751.3 | Intron | NP_003742.2 | ||||
XM_011515599.1 | Intron | XP_011513901.1 | ||||
XM_011515600.1 | Intron | XP_011513902.1 | ||||
XM_011515601.1 | Intron | XP_011513903.1 | ||||
XM_017012752.1 | Intron | XP_016868241.1 | ||||
XM_017012753.1 | Intron | XP_016868242.1 |
SNX8 - sorting nexin 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |