Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATGAAAATGACTTAAATTAAGGG[A/T]TGAATTAATTGTGTAAACATATAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142955 MIM: 604685 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HOTAIRM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HOTAIRM1 - HOXA transcript antisense RNA, myeloid-specific 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXA1 - homeobox A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005522.4 | 1369 | UTR 3 | NP_005513.1 | |||
NM_153620.2 | 1369 | UTR 3 | NP_705873.2 |
HOXA2 - homeobox A2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |