Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTGAACTTTGTGCTGGTGACTTT[C/T]AATTATGGTGTTCTCCTGGCCCGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609225 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
WIPI2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WIPI2 - WD repeat domain, phosphoinositide interacting 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033518.1 | Intron | NP_001028690.1 | ||||
NM_001033519.1 | Intron | NP_001028691.1 | ||||
NM_001033520.1 | Intron | NP_001028692.1 | ||||
NM_001278299.1 | Intron | NP_001265228.1 | ||||
NM_015610.3 | Intron | NP_056425.1 | ||||
NM_016003.3 | Intron | NP_057087.2 | ||||
XM_006715685.1 | Intron | XP_006715748.1 | ||||
XM_017011952.1 | Intron | XP_016867441.1 |