Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGCTGGGTTGTTGCTGGTTTTGT[G/T]GTGGCCTCTTCCGGTCCTTGATCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609603 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRYGN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CRYGN - crystallin gamma N | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308292.1 | 765 | UTR 3 | NP_001295221.1 | |||
NM_144727.2 | 765 | Silent Mutation | ACA,ACC | T,T 167 | NP_653328.1 | |
XM_005249952.3 | 765 | UTR 3 | XP_005250009.1 | |||
XM_011515836.1 | 765 | Intron | XP_011514138.1 | |||
XM_011515837.2 | 765 | Intron | XP_011514139.1 | |||
XM_017011773.1 | 765 | Intron | XP_016867262.1 |
MIR3907 - microRNA 3907 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |