Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGGGCGACTGGAGCCATGGCGGA[G/T]TCGGCGCCTGCTCGGGTAAAGAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602386 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF212 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF212 - zinc finger protein 212 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012256.3 | 140 | Missense Mutation | GAG,GAT | E,D 3 | NP_036388.2 | |
XM_005250055.2 | 140 | Missense Mutation | AGT,ATT | S,I 45 | XP_005250112.2 | |
XM_011516581.2 | 140 | UTR 5 | XP_011514883.2 | |||
XM_011516582.1 | 140 | Intron | XP_011514884.1 | |||
XM_017012646.1 | 140 | Missense Mutation | GAG,GAT | E,D 3 | XP_016868135.1 | |
XM_017012647.1 | 140 | UTR 5 | XP_016868136.1 | |||
XM_017012648.1 | 140 | UTR 5 | XP_016868137.1 |