Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGGACAGAAGACAACAGCTAAAA[C/T]AGAGAAAGGACTTCAGAAATGTGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601677 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASB15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ASB15 - ankyrin repeat and SOCS box containing 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290258.1 | Intron | NP_001277187.1 | ||||
NM_080928.4 | Intron | NP_563616.3 | ||||
XM_005250146.4 | Intron | XP_005250203.1 | ||||
XM_011515816.2 | Intron | XP_011514118.1 | ||||
XM_011515817.2 | Intron | XP_011514119.1 | ||||
XM_011515818.2 | Intron | XP_011514120.1 | ||||
XM_011515819.2 | Intron | XP_011514121.1 | ||||
XM_011515820.2 | Intron | XP_011514122.1 | ||||
XM_011515821.2 | Intron | XP_011514123.1 | ||||
XM_017011756.1 | Intron | XP_016867245.1 | ||||
XM_017011757.1 | Intron | XP_016867246.1 | ||||
XM_017011758.1 | Intron | XP_016867247.1 |
NDUFA5 - NADH:ubiquinone oxidoreductase subunit A5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282419.2 | Intron | NP_001269348.1 | ||||
NM_001282420.2 | Intron | NP_001269349.1 | ||||
NM_001282421.2 | Intron | NP_001269350.1 | ||||
NM_001282422.2 | Intron | NP_001269351.1 | ||||
NM_001291304.1 | Intron | NP_001278233.1 | ||||
NM_005000.4 | Intron | NP_004991.1 |