Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTCTGGATGCTTTTCTGTGGGAT[A/G]GGGGAGGGGGCATCCACTCTGATTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603420 MIM: 613522 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CALU PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CALU - calumenin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130674.2 | Intron | NP_001124146.1 | ||||
NM_001199671.1 | Intron | NP_001186600.1 | ||||
NM_001199672.1 | Intron | NP_001186601.1 | ||||
NM_001199673.1 | Intron | NP_001186602.1 | ||||
NM_001219.4 | Intron | NP_001210.1 | ||||
XM_017012659.1 | Intron | XP_016868148.1 |
OPN1SW - opsin 1 (cone pigments), short-wave-sensitive | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001708.2 | Intron | NP_001699.1 |