Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGAACTTTCTTACAGAGAAAAAT[A/C]TTTTTTCCTTAAAAATGTATATGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610523 MIM: 114850 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP41 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CEP41 - centrosomal protein 41 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257158.1 | 3186 | UTR 3 | NP_001244087.1 | |||
NM_001257159.1 | 3186 | UTR 3 | NP_001244088.1 | |||
NM_001257160.1 | 3186 | Intron | NP_001244089.1 | |||
NM_018718.2 | 3186 | UTR 3 | NP_061188.1 | |||
XM_011516709.2 | 3186 | UTR 3 | XP_011515011.1 | |||
XM_011516710.2 | 3186 | UTR 3 | XP_011515012.1 |
CPA1 - carboxypeptidase A1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |