Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCCTCAAGTACATACAAACATGA[A/G]GGCCGGTAGACAAAATATATCCTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603961 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEMA3A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SEMA3A - semaphorin 3A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006080.2 | 3127 | UTR 3 | NP_006071.1 | |||
XM_005250110.3 | 3127 | UTR 3 | XP_005250167.1 | |||
XM_005250111.4 | 3127 | UTR 3 | XP_005250168.1 | |||
XM_006715839.3 | 3127 | UTR 3 | XP_006715902.1 | |||
XM_011515734.2 | 3127 | UTR 3 | XP_011514036.1 | |||
XM_017011673.1 | 3127 | UTR 3 | XP_016867162.1 |