Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGACAGTTACCTTTGCTCTTCC[A/G]CAGGAGCTTGTGTGTTTTCTCTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607773 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLEKHA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLEKHA2 - pleckstrin homology domain containing A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021623.1 | Intron | NP_067636.1 | ||||
XM_011544605.2 | Intron | XP_011542907.1 | ||||
XM_011544606.2 | Intron | XP_011542908.1 | ||||
XM_011544607.2 | Intron | XP_011542909.1 | ||||
XM_017013718.1 | Intron | XP_016869207.1 | ||||
XM_017013719.1 | Intron | XP_016869208.1 | ||||
XM_017013720.1 | Intron | XP_016869209.1 |