Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTAAGAAGAATTCTCTCCTGTTC[C/T]AAGGCTTCTATATACTTTGAGTAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615650 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RGS22 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RGS22 - regulator of G-protein signaling 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286692.1 | 4039 | Silent Mutation | TTA,TTG | L,L 1205 | NP_001273621.1 | |
NM_001286693.1 | 4039 | Silent Mutation | TTA,TTG | L,L 1036 | NP_001273622.1 | |
NM_015668.4 | 4039 | Silent Mutation | TTA,TTG | L,L 1217 | NP_056483.3 | |
XM_005250856.2 | 4039 | Silent Mutation | TTA,TTG | L,L 1164 | XP_005250913.1 | |
XM_005250857.2 | 4039 | Silent Mutation | TTA,TTG | L,L 1164 | XP_005250914.1 | |
XM_005250858.2 | 4039 | Silent Mutation | TTA,TTG | L,L 1163 | XP_005250915.1 | |
XM_005250860.2 | 4039 | Intron | XP_005250917.1 | |||
XM_006716540.2 | 4039 | Silent Mutation | TTA,TTG | L,L 1121 | XP_006716603.1 | |
XM_011516957.1 | 4039 | Silent Mutation | TTA,TTG | L,L 1121 | XP_011515259.1 | |
XM_011516958.1 | 4039 | Silent Mutation | TTA,TTG | L,L 1121 | XP_011515260.1 | |
XM_011516959.2 | 4039 | Intron | XP_011515261.1 | |||
XM_017013309.1 | 4039 | Silent Mutation | TTA,TTG | L,L 1111 | XP_016868798.1 | |
XM_017013310.1 | 4039 | Silent Mutation | TTA,TTG | L,L 1110 | XP_016868799.1 | |
XM_017013311.1 | 4039 | Silent Mutation | TTA,TTG | L,L 762 | XP_016868800.1 |