Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTTCTATCAGCTTCCTTTTCTTT[A/T]TCTTGTTCCTCTTCTCCTTCGCCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610575 | ||||||||||||||||||||
Literature Links: |
RSPO2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RSPO2 - R-spondin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282863.1 | 1300 | Missense Mutation | AAA,ATA | K,I 155 | NP_001269792.1 | |
NM_001317942.1 | 1300 | Missense Mutation | AAA,ATA | K,I 152 | NP_001304871.1 | |
NM_178565.4 | 1300 | Missense Mutation | AAA,ATA | K,I 219 | NP_848660.3 | |
XM_011517018.1 | 1300 | Missense Mutation | AAA,ATA | K,I 226 | XP_011515320.1 | |
XM_011517019.1 | 1300 | Missense Mutation | AAA,ATA | K,I 225 | XP_011515321.1 | |
XM_017013395.1 | 1300 | Missense Mutation | AAA,ATA | K,I 218 | XP_016868884.1 |