Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCTGGGATACAGGCTTGAGCCAC[C/T]GCACCTGACCTTGGCATGTACTTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609884 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM67 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM67 - transmembrane protein 67 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142301.1 | 352 | Missense Mutation | CCG,CTG | P,L 36 | NP_001135773.1 | |
NM_153704.5 | 352 | Intron | NP_714915.3 | |||
XM_006716686.3 | 352 | Intron | XP_006716749.1 | |||
XM_011517363.2 | 352 | Intron | XP_011515665.1 |