Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGTCTCATGTTTGGTTTAAAGCA[A/C]TTCCAAAATATCAGACAGTTATGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613945 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJC5B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAJC5B - DnaJ heat shock protein family (Hsp40) member C5 beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033105.4 | Intron | NP_149096.2 | ||||
XM_011517618.2 | Intron | XP_011515920.1 | ||||
XM_011517619.2 | Intron | XP_011515921.1 | ||||
XM_011517620.2 | Intron | XP_011515922.1 | ||||
XM_011517621.2 | Intron | XP_011515923.1 |