Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAACACCTGGATCTGACAATTTAAT[A/G]TTTTAAGTATTTTGAATCTTTGTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605966 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HNF4G PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HNF4G - hepatocyte nuclear factor 4 gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004133.4 | Intron | NP_004124.4 | ||||
XM_011517516.2 | Intron | XP_011515818.1 | ||||
XM_017013373.1 | Intron | XP_016868862.1 | ||||
XM_017013374.1 | Intron | XP_016868863.1 | ||||
XM_017013375.1 | Intron | XP_016868864.1 | ||||
XM_017013376.1 | Intron | XP_016868865.1 |