Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCTTTTCCTCCAAAGCTCCTATT[A/G]TTTGACATGCCACTTCTTGCTTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607340 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GABBR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GABBR2 - gamma-aminobutyric acid type B receptor subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005458.7 | Intron | NP_005449.5 | ||||
XM_005252316.4 | Intron | XP_005252373.1 | ||||
XM_017015331.1 | Intron | XP_016870820.1 | ||||
XM_017015332.1 | Intron | XP_016870821.1 |